Assay Details
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024285.1 | NP_001019456.1 | 1 - 2 | 73 | 65 | |
GenBank mRNA | AB174900.2 | - | 1 - 2 | 73 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB174900.2, NM_001024285.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
