Assay Details
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024285.1 | NP_001019456.1 | 5 - 6 | 516 | 87 | |
GenBank mRNA | AB174900.2 | - | 5 - 6 | 516 | 87 | |
AY367561.1 | - | 5 - 6 | 516 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
