Assay Details
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024285.1 | NP_001019456.1 | 7 - 8 | 778 | 153 | |
GenBank mRNA | AB174900.2 | - | 7 - 8 | 778 | 153 | |
AY367561.1 | - | 7 - 8 | 778 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
signal transduction
response to xenobiotic stimulus
positive regulation of protein sumoylation
negative regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
DNA binding
protein dimerization activity
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
signal transduction
response to xenobiotic stimulus
positive regulation of protein sumoylation
negative regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
DNA binding
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P011324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75809 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P011324 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P011324 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P011324 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28255 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50141 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69441 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49133 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11198 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10702504 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702493 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702496 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702502 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702498 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702501 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702500 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702503 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702499 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702497 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702494 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702495 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10702492 | - | - |
Rat Genome 230 2.0 Array | 1380746_PM_at | - | - |
Rat Genome 230 2.0 Array | 1380746_at | - | - |
Affymetrix HT Rat Focus Array | 1380746_at | - | - |
Affymetrix HT Rat Focus Array | 1380746_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100001898.rn.1 | TC0100000330.rn.1 | RTA TC |
Rat Expression Set 230 | 1380746_PM_at | - | - |
Rat Expression Set 230 | 1380746_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702492 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22007488 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368540 | - | - |
