Assay Details
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193568.1 | NP_001180497.1 | 6 - 7 | 694 | 119 | |
NM_001193569.1 | NP_001180498.1 | 6 - 7 | 632 | 119 | ||
NM_019232.3 | NP_062105.2 | 6 - 7 | 606 | 119 | ||
XM_006227723.1 | XP_006227785.1 | 8 - 9 | 1292 | 119 | ||
XM_008758606.1 | XP_008756828.1 | 6 - 7 | 802 | 119 | ||
GenBank mRNA | BC078843.1 | - | 6 - 7 | 560 | 119 | |
FM068981.1 | - | 6 - 7 | 612 | 119 | ||
L01624.1 | - | 6 - 7 | 611 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
