Assay Details
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193568.1 | NP_001180497.1 | 7 - 8 | 803 | 108 | |
NM_001193569.1 | NP_001180498.1 | 7 - 8 | 741 | 108 | ||
NM_019232.3 | NP_062105.2 | 7 - 8 | 715 | 108 | ||
XM_006227723.1 | XP_006227785.1 | 9 - 10 | 1401 | 108 | ||
XM_008758606.1 | XP_008756828.1 | 7 - 8 | 911 | 108 | ||
GenBank mRNA | BC078843.1 | - | 7 - 8 | 669 | 108 | |
L01624.1 | - | 7 - 8 | 720 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
