Assay Details
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021869.2 | NP_068641.2 | 2 - 3 | 182 | 151 | |
XM_006227691.2 | XP_006227753.1 | 3 - 4 | 1504 | 151 | ||
XM_008758626.1 | XP_008756848.1 | 3 - 4 | 1498 | 151 | ||
GenBank mRNA | AF031430.1 | - | 1 - 2 | 86 | 151 | |
BC085737.1 | - | 2 - 3 | 182 | 7193349 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
