Assay Details
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021869.2 | NP_068641.2 | 7 - 8 | 634 | 134 | |
XM_006227691.2 | XP_006227753.1 | 8 - 9 | 1956 | 134 | ||
XM_008758626.1 | XP_008756848.1 | 8 - 9 | 1950 | 134 | ||
GenBank mRNA | AF031430.1 | - | 6 - 7 | 538 | 134 | |
BC085737.1 | - | 7 - 8 | 634 | 134 |
Gene Symbol: LOC100910446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758670.1 | XP_008756892.1 | 5 - 6 | 520 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
