Assay Details
Gene Symbol: Srd5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017070.3 | NP_058766.2 | 704 | 86 | ||
GenBank mRNA | BC127454.1 | - | 705 | 86 | ||
EF091857.1 | - | 190 | 86 | |||
J05035.1 | - | 557 | 86 | |||
S81448.1 | - | 577 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
