Assay Details
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_220095.7 | XP_220095.4 | 1 - 2 | 325 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
