Assay Details
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_220095.7 | XP_220095.4 | 4 - 5 | 721 | 97 | |
GenBank mRNA | BC091331.1 | - | 2 - 3 | 105 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
