Assay Details
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_220095.7 | XP_220095.4 | 6 - 7 | 1000 | 91 | |
GenBank mRNA | BC091331.1 | - | 4 - 5 | 384 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
