Assay Details
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_220095.7 | XP_220095.4 | 7 - 8 | 1171 | 76 | |
GenBank mRNA | BC091331.1 | - | 5 - 6 | 555 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
