Assay Details
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130428.1 | NP_569112.1 | 10 - 11 | 1451 | 62 | |
GenBank mRNA | AB072907.1 | - | 10 - 11 | 1451 | 62 | |
DQ402976.1 | - | 8 - 9 | 1260 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
