Assay Details
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130428.1 | NP_569112.1 | 5 - 6 | 640 | 88 | |
GenBank mRNA | AB072907.1 | - | 5 - 6 | 640 | 88 | |
DQ402976.1 | - | 3 - 4 | 449 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
