Assay Details
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130428.1 | NP_569112.1 | 9 - 10 | 1272 | 97 | |
GenBank mRNA | AB072907.1 | - | 9 - 10 | 1272 | 97 | |
DQ402976.1 | - | 7 - 8 | 1081 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
