Assay Details
Gene Symbol: Echdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007734.1 | NP_001007735.1 | 2 - 3 | 351 | 117 | |
XM_006227746.2 | XP_006227808.1 | 2 - 3 | 541 | 117 | ||
XM_006227747.2 | XP_006227809.1 | 2 - 3 | 1138 | 117 | ||
GenBank mRNA | BC079052.1 | - | 2 - 3 | 351 | 7115386 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
