Assay Details
Gene Symbol: Echdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007734.1 | NP_001007735.1 | 4 - 5 | 549 | 77 | |
XM_006227746.2 | XP_006227808.1 | 4 - 5 | 739 | 77 | ||
XM_006227747.2 | XP_006227809.1 | 4 - 5 | 1336 | 77 | ||
GenBank mRNA | BC079052.1 | - | 4 - 5 | 549 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
