Assay Details
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007620.1 | NP_001007621.1 | 1 - 2 | 367 | 92 | |
XM_006251754.1 | XP_006251816.1 | 2 - 3 | 227 | 92 | ||
GenBank mRNA | BC079137.1 | - | 1 - 2 | 367 | 7127701 | 92 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079137.1,NM_001007620.1,XM_006251754.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
