Assay Details
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270854.1 | NP_001257783.1 | 9 - 10 | 1135 | 76 | |
NM_012672.3 | NP_036804.2 | 10 - 11 | 1243 | 76 | ||
XM_006251709.2 | XP_006251771.1 | 5 - 6 | 1156 | 76 | ||
XM_008770500.1 | XP_008768722.1 | 7 - 8 | 1047 | 76 | ||
GenBank mRNA | DQ191165.1 | - | 7 - 8 | 990 | 76 | |
HM043807.1 | - | 6 - 7 | 1032 | 76 | ||
J03819.1 | - | 9 - 10 | 1131 | 76 | ||
J03933.1 | - | 9 - 10 | 1132 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
