Assay Details
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270854.1 | NP_001257783.1 | 3 - 4 | 214 | 67 | |
NM_012672.3 | NP_036804.2 | 3 - 4 | 214 | 67 | ||
GenBank mRNA | J03819.1 | - | 3 - 4 | 210 | 67 | |
J03933.1 | - | 3 - 4 | 211 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the J03819.1,J03933.1,NM_001270854.1,NM_012672.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
