Assay Details
Gene Symbol: Rarb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031529.1 | NP_113717.1 | 2 - 3 | 841 | 86 | |
GenBank mRNA | AJ002942.1 | - | 2 - 3 | 499 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
