Assay Details
Gene Symbol: LOC100911261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751445.3 | XP_003751493.2 | 1015 | 82 | ||
XM_008770601.1 | XP_008768823.1 | 1015 | 82 | |||
XM_008770602.1 | XP_008768824.1 | 1015 | 82 |
Gene Symbol: Flnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107288.1 | NP_001100758.1 | 26 - 27 | 4643 | 82 | |
XM_006251780.2 | XP_006251842.1 | 26 - 27 | 4704 | 82 | ||
XM_006251781.2 | XP_006251843.1 | 26 - 27 | 4704 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
