Assay Details
Gene Symbol: LOC100911261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751445.3 | XP_003751493.2 | 3657 | 100 | ||
XM_008770601.1 | XP_008768823.1 | 3690 | 100 | |||
XM_008770602.1 | XP_008768824.1 | 3618 | 100 |
Gene Symbol: Flnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107288.1 | NP_001100758.1 | 42 - 43 | 7246 | 100 | |
XM_006251780.2 | XP_006251842.1 | 43 - 44 | 7379 | 100 | ||
XM_006251781.2 | XP_006251843.1 | 43 - 44 | 7346 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P513145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14297 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P513145 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P513145 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P513145 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P295151 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11945 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77511 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5703 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71042 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10782706 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782716 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782705 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782724 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782747 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782726 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782711 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782741 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782751 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782712 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782745 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782743 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782750 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782744 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782715 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782708 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782748 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782721 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782734 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782725 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782720 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782707 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782729 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782710 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782718 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782733 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782749 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782717 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782723 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782730 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782735 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782719 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782713 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782746 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782714 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10782704 | - | - |
Rat Genome 230 2.0 Array | 1388401_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388401_at | - | - |
Rat Genome 230 2.0 Array | 1391827_at | - | - |
Rat Genome 230 2.0 Array | 1391827_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391827_at | - | - |
Affymetrix HT Rat Focus Array | 1391827_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388401_at | - | - |
Affymetrix HT Rat Focus Array | 1388401_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500009040.rn.1 | TC1500001420.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500009041.rn.1 | TC1500001420.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500009039.rn.1 | TC1500001420.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500009043.rn.1 | TC1500001420.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500009042.rn.1 | TC1500001420.rn.1 | RTA TC |
Rat Expression Set 230 | 1388401_PM_at | - | - |
Rat Expression Set 230 | 1388401_at | - | - |
Rat Expression Set 230 | 1391827_PM_at | - | - |
Rat Expression Set 230 | 1391827_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10782704 | - | - |
Rat Genome U34 Set | rc_AA892820_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21886362 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362723 | - | - |
