Assay Details
Gene Symbol: Papd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107416.1 | NP_001100886.1 | 1 - 2 | 208 | 150 | |
XM_006255261.1 | XP_006255323.1 | 1 - 2 | 614 | 150 | ||
XM_006255263.2 | XP_006255325.1 | 1 - 2 | 614 | 150 | ||
XM_008772403.1 | XP_008770625.1 | 2 - 3 | 625 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107416.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
