Assay Details
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012057.1 | NP_001012057.1 | 5 - 6 | 844 | 77 | |
XM_008772405.1 | XP_008770627.1 | 6 - 7 | 965 | 77 | ||
XM_008772406.1 | XP_008770628.1 | 6 - 7 | 1154 | 77 | ||
XM_008772407.1 | XP_008770629.1 | 6 - 7 | 891 | 77 | ||
GenBank mRNA | FQ223845.1 | - | 1040 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
