Assay Details
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012057.1 | NP_001012057.1 | 6 - 7 | 919 | 68 | |
XM_008772405.1 | XP_008770627.1 | 7 - 8 | 1040 | 68 | ||
XM_008772406.1 | XP_008770628.1 | 7 - 8 | 1229 | 68 | ||
XM_008772407.1 | XP_008770629.1 | 7 - 8 | 966 | 68 | ||
GenBank mRNA | FQ223845.1 | - | 1115 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
