Assay Details
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011926.1 | NP_001011926.1 | 2 - 3 | 207 | 73 | |
XM_006255181.2 | XP_006255243.1 | 2 - 3 | 219 | 73 | ||
XM_006255182.2 | XP_006255244.1 | 2 - 3 | 197 | 73 | ||
XM_006255183.2 | XP_006255245.1 | 1 - 2 | 120 | 73 | ||
XM_006255185.2 | XP_006255247.1 | 2 - 3 | 153 | 73 | ||
GenBank mRNA | BC089985.1 | - | 2 - 3 | 207 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
