Assay Details
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011926.1 | NP_001011926.1 | 4 - 5 | 476 | 88 | |
XM_006255181.2 | XP_006255243.1 | 4 - 5 | 488 | 88 | ||
XM_006255182.2 | XP_006255244.1 | 4 - 5 | 466 | 88 | ||
XM_006255183.2 | XP_006255245.1 | 3 - 4 | 389 | 88 | ||
XM_006255185.2 | XP_006255247.1 | 4 - 5 | 422 | 88 | ||
GenBank mRNA | BC089985.1 | - | 4 - 5 | 476 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
