Assay Details
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011926.1 | NP_001011926.1 | 5 - 6 | 571 | 101 | |
XM_006255181.2 | XP_006255243.1 | 5 - 6 | 583 | 101 | ||
XM_006255182.2 | XP_006255244.1 | 5 - 6 | 561 | 101 | ||
XM_006255183.2 | XP_006255245.1 | 4 - 5 | 484 | 101 | ||
XM_006255185.2 | XP_006255247.1 | 5 - 6 | 517 | 101 | ||
GenBank mRNA | BC089985.1 | - | 5 - 6 | 571 | 7318380 | 101 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
