Assay Details
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011926.1 | NP_001011926.1 | 7 - 8 | 756 | 135 | |
XM_006255181.2 | XP_006255243.1 | 7 - 8 | 768 | 135 | ||
XM_006255182.2 | XP_006255244.1 | 7 - 8 | 746 | 135 | ||
XM_006255183.2 | XP_006255245.1 | 6 - 7 | 669 | 135 | ||
XM_006255185.2 | XP_006255247.1 | 7 - 8 | 702 | 135 | ||
GenBank mRNA | BC089985.1 | - | 7 - 8 | 756 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
