Assay Details
Gene Symbol: Ggps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771698.1 | XP_008769920.1 | 4 - 5 | 955 | 133 | |
GenBank mRNA | AB118241.1 | - | 4 - 5 | 923 | 133 | |
AB118242.1 | - | 4 - 5 | 923 | 133 | ||
AB118243.1 | - | 4 - 5 | 923 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
