Assay Details
Gene Symbol: Zeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U51583.1 | - | 248 | 62 | ||
U51584.1 | - | 1389 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U51584.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
