Assay Details
Gene Symbol: Zeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013164.1 | NP_037296.1 | 6 - 7 | 2272 | 72 | |
GenBank mRNA | AY325179.1 | - | 6 - 7 | 2272 | 72 | |
U51583.1 | - | 2519 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
