Assay Details
Gene Symbol: Nexn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139230.1 | NP_631976.1 | 8 - 9 | 1184 | 158 | |
NM_139231.1 | NP_631977.1 | 8 - 9 | 1034 | 158 | ||
XM_006233479.2 | XP_006233541.1 | 10 - 11 | 1352 | 158 | ||
XM_006233480.2 | XP_006233542.1 | 10 - 11 | 1601 | 158 | ||
XM_006233483.2 | XP_006233545.1 | 9 - 10 | 1310 | 158 | ||
XM_006233484.2 | XP_006233546.1 | 9 - 10 | 1159 | 158 | ||
XM_006233485.2 | XP_006233547.1 | 8 - 9 | 1117 | 158 | ||
XM_008761548.1 | XP_008759770.1 | 9 - 10 | 1301 | 158 | ||
GenBank mRNA | AF056034.1 | - | 8 - 9 | 1184 | 158 | |
AF056035.1 | - | 8 - 9 | 1034 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
