Assay Details
Gene Symbol: Oaz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109899.2 | NP_001103369.1 | 4 - 5 | 611 | 74 | |
NM_001301297.1 | NP_001288226.1 | 4 - 5 | 608 | 74 | ||
GenBank mRNA | BI290904.1 | - | 1 - 2 | 74 | ||
FQ211793.1 | - | 4 - 5 | 611 | 74 | ||
FQ214772.1 | - | 4 - 5 | 611 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
