Assay Details
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012496.2 | NP_036628.2 | 3 - 4 | 385 | 105 | |
GenBank mRNA | BC081697.1 | - | 3 - 4 | 385 | 105 | |
FQ209505.1 | - | 3 - 4 | 405 | 105 | ||
FQ209538.1 | - | 3 - 4 | 405 | 105 | ||
FQ209583.1 | - | 3 - 4 | 405 | 105 | ||
FQ209594.1 | - | 3 - 4 | 405 | 105 | ||
FQ209615.1 | - | 3 - 4 | 105 | |||
FQ209621.1 | - | 3 - 4 | 403 | 105 | ||
FQ209639.1 | - | 3 - 4 | 405 | 105 | ||
FQ209643.1 | - | 3 - 4 | 402 | 105 | ||
FQ209782.1 | - | 3 - 4 | 405 | 105 | ||
FQ209819.1 | - | 3 - 4 | 415 | 105 | ||
FQ210002.1 | - | 3 - 4 | 405 | 105 | ||
FQ210453.1 | - | 1 - 2 | 112 | 105 | ||
FQ210849.1 | - | 3 - 4 | 405 | 105 | ||
FQ211083.1 | - | 3 - 4 | 405 | 105 | ||
FQ211156.1 | - | 3 - 4 | 405 | 105 | ||
FQ211159.1 | - | 3 - 4 | 405 | 105 | ||
FQ211293.1 | - | 1 - 2 | 83 | 105 | ||
FQ218139.1 | - | 3 - 4 | 405 | 105 | ||
FQ218217.1 | - | 3 - 4 | 405 | 105 | ||
FQ218348.1 | - | 3 - 4 | 411 | 105 | ||
FQ218402.1 | - | 3 - 4 | 407 | 105 | ||
FQ218551.1 | - | 3 - 4 | 405 | 105 | ||
FQ218665.1 | - | 3 - 4 | 405 | 105 | ||
FQ218885.1 | - | 3 - 4 | 405 | 105 | ||
FQ218911.1 | - | 3 - 4 | 405 | 105 | ||
FQ219077.1 | - | 1 - 2 | 89 | 105 | ||
FQ219096.1 | - | 3 - 4 | 405 | 105 | ||
FQ219116.1 | - | 3 - 4 | 405 | 105 | ||
FQ219236.1 | - | 3 - 4 | 404 | 105 | ||
FQ219261.1 | - | 3 - 4 | 405 | 105 | ||
FQ219453.1 | - | 3 - 4 | 405 | 105 | ||
FQ219555.1 | - | 3 - 4 | 405 | 105 | ||
FQ219585.1 | - | 3 - 4 | 405 | 105 | ||
FQ219640.1 | - | 3 - 4 | 403 | 105 | ||
FQ219646.1 | - | 3 - 4 | 405 | 105 | ||
FQ219703.1 | - | 3 - 4 | 404 | 105 | ||
M10149.1 | - | 3 - 4 | 396 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
