Assay Details
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012496.2 | NP_036628.2 | 4 - 5 | 436 | 87 | |
GenBank mRNA | BC081697.1 | - | 4 - 5 | 436 | 87 | |
FQ209505.1 | - | 4 - 5 | 456 | 87 | ||
FQ209538.1 | - | 4 - 5 | 456 | 87 | ||
FQ209583.1 | - | 4 - 5 | 456 | 87 | ||
FQ209594.1 | - | 4 - 5 | 456 | 87 | ||
FQ209615.1 | - | 4 - 5 | 456 | 87 | ||
FQ209621.1 | - | 4 - 5 | 454 | 87 | ||
FQ209639.1 | - | 4 - 5 | 456 | 87 | ||
FQ209643.1 | - | 4 - 5 | 453 | 87 | ||
FQ209782.1 | - | 4 - 5 | 456 | 87 | ||
FQ209819.1 | - | 4 - 5 | 466 | 87 | ||
FQ210002.1 | - | 4 - 5 | 456 | 87 | ||
FQ210453.1 | - | 2 - 3 | 163 | 87 | ||
FQ210849.1 | - | 4 - 5 | 456 | 87 | ||
FQ211083.1 | - | 4 - 5 | 456 | 87 | ||
FQ211156.1 | - | 4 - 5 | 456 | 87 | ||
FQ211159.1 | - | 4 - 5 | 456 | 87 | ||
FQ211293.1 | - | 2 - 3 | 134 | 87 | ||
FQ218139.1 | - | 4 - 5 | 456 | 87 | ||
FQ218217.1 | - | 4 - 5 | 456 | 87 | ||
FQ218348.1 | - | 4 - 5 | 462 | 87 | ||
FQ218402.1 | - | 4 - 5 | 458 | 87 | ||
FQ218522.1 | - | 4 - 5 | 456 | 87 | ||
FQ218551.1 | - | 4 - 5 | 456 | 87 | ||
FQ218665.1 | - | 4 - 5 | 456 | 87 | ||
FQ218885.1 | - | 4 - 5 | 456 | 87 | ||
FQ218911.1 | - | 4 - 5 | 456 | 87 | ||
FQ219077.1 | - | 2 - 3 | 140 | 87 | ||
FQ219096.1 | - | 4 - 5 | 456 | 87 | ||
FQ219116.1 | - | 4 - 5 | 456 | 87 | ||
FQ219236.1 | - | 4 - 5 | 455 | 87 | ||
FQ219261.1 | - | 4 - 5 | 456 | 87 | ||
FQ219453.1 | - | 4 - 5 | 456 | 87 | ||
FQ219555.1 | - | 4 - 5 | 456 | 87 | ||
FQ219585.1 | - | 4 - 5 | 456 | 87 | ||
FQ219640.1 | - | 4 - 5 | 454 | 87 | ||
FQ219646.1 | - | 4 - 5 | 456 | 87 | ||
FQ219703.1 | - | 4 - 5 | 455 | 87 | ||
M10149.1 | - | 4 - 5 | 447 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
