Assay Details
Gene Symbol: Glud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012570.2 | NP_036702.1 | 10 - 11 | 1571 | 171 | |
GenBank mRNA | BC081841.1 | - | 10 - 11 | 1455 | 171 | |
FQ214067.1 | - | 2 - 3 | 151 | 171 | ||
U95148.1 | - | 1 - 2 | 112 | 171 | ||
X14044.1 | - | 10 - 11 | 1461 | 171 | ||
X14223.1 | - | 10 - 11 | 1434 | 171 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
