Assay Details
Gene Symbol: Ivd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012592.2 | NP_036724.1 | 5 - 6 | 558 | 68 | |
GenBank mRNA | BC088401.1 | - | 5 - 6 | 558 | 68 | |
FQ231497.1 | - | 4 - 5 | 400 | 68 | ||
J05031.1 | - | 5 - 6 | 568 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
