Assay Details
Gene Symbol: Me1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012600.2 | NP_036732.2 | 13 - 14 | 1550 | 106 | |
XM_008766439.1 | XP_008764661.1 | 13 - 14 | 1847 | 106 | ||
GenBank mRNA | FQ228526.1 | - | 13 - 14 | 1581 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
