Assay Details
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012895.3 | NP_037027.2 | 9 - 10 | 890 | 91 | |
XM_006251631.1 | XP_006251693.1 | 9 - 10 | 1004 | 91 | ||
GenBank mRNA | BC081712.1 | - | 9 - 10 | 890 | 91 | |
FQ209450.1 | - | 4 - 5 | 359 | 91 | ||
FQ219573.1 | - | 3 - 4 | 198 | 91 | ||
FQ225093.1 | - | 5 - 6 | 561 | 91 | ||
U57042.1 | - | 8 - 9 | 830 | 91 | ||
U90340.1 | - | 9 - 10 | 91 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
