Assay Details
Gene Symbol: Cpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013128.1 | NP_037260.1 | 8 - 9 | 1419 | 90 | |
GenBank mRNA | BC085762.1 | - | 8 - 9 | 1418 | 90 | |
J04625.1 | - | 8 - 9 | 1374 | 90 | ||
M31602.1 | - | 8 - 9 | 1390 | 90 | ||
X51406.1 | - | 8 - 9 | 1419 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
