Assay Details
Gene Symbol: Rheb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013216.1 | NP_037348.1 | 5 - 6 | 455 | 71 | |
GenBank mRNA | BC088857.1 | - | 5 - 6 | 509 | 7307969 | 71 |
FQ212847.1 | - | 5 - 6 | 477 | 71 | ||
FQ214530.1 | - | 5 - 6 | 494 | 71 | ||
FQ218725.1 | - | 5 - 6 | 477 | 71 | ||
FQ225701.1 | - | 5 - 6 | 459 | 71 | ||
FQ230203.1 | - | 5 - 6 | 459 | 71 | ||
U08227.1 | - | 5 - 6 | 455 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
