Assay Details
Gene Symbol: Ahcy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017201.1 | NP_058897.1 | 1 - 2 | 55 | 95 | |
GenBank mRNA | M15185.1 | - | 1 - 2 | 55 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M15185.1, NM_017201.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
