Assay Details
Gene Symbol: Gclm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017305.2 | NP_059001.1 | 1 - 2 | 248 | 80 | |
GenBank mRNA | BC078867.1 | - | 1 - 2 | 248 | 80 | |
L22191.1 | - | 1 - 2 | 186 | 80 | ||
S65555.1 | - | 1 - 2 | 186 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
