Assay Details
Gene Symbol: Gclm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017305.2 | NP_059001.1 | 5 - 6 | 656 | 89 | |
GenBank mRNA | BC078867.1 | - | 5 - 6 | 656 | 89 | |
DQ266366.1 | - | 4 - 5 | 398 | 89 | ||
L22191.1 | - | 5 - 6 | 594 | 89 | ||
S65555.1 | - | 5 - 6 | 594 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
