Assay Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019238.2 | NP_062111.1 | 1 - 2 | 175 | 89 | |
GenBank mRNA | BC081810.1 | - | 1 - 2 | 175 | 89 | |
FQ213323.1 | - | 1 - 2 | 158 | 89 | ||
FQ219372.1 | - | 1 - 2 | 146 | 89 | ||
M95591.1 | - | 1 - 2 | 193 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
