Assay Details
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195505.1 | NP_001182434.1 | 463 | 78 | ||
NM_001195506.1 | NP_001182435.1 | 334 | 78 | |||
NM_019257.3 | NP_062130.2 | 338 | 78 | |||
XM_008764878.1 | XP_008763100.1 | 3 - 4 | 514 | 78 | ||
XM_008764879.1 | XP_008763101.1 | 3 - 4 | 385 | 78 | ||
GenBank mRNA | AF020683.1 | - | 3 - 4 | 78 | ||
BC058479.1 | - | 332 | 6920786 | 78 | ||
FM049719.1 | - | 3 - 4 | 426 | 78 | ||
FM059586.1 | - | 3 - 4 | 312 | 78 | ||
FM081441.1 | - | 3 - 4 | 338 | 78 | ||
L13635.1 | - | 434 | 78 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
