Assay Details
Gene Symbol: Prkar2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019264.2 | NP_062137.1 | 4 - 5 | 540 | 85 | |
XM_006243712.2 | XP_006243774.1 | 5 - 6 | 725 | 85 | ||
XM_006243713.2 | XP_006243775.1 | 5 - 6 | 539 | 85 | ||
XM_008766478.1 | XP_008764700.1 | 4 - 5 | 490 | 85 | ||
GenBank mRNA | AF533978.1 | - | 4 - 5 | 544 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
