Assay Details
Gene Symbol: Vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021847.3 | NP_068619.1 | 4 - 5 | 398 | 157 | |
XM_006235706.2 | XP_006235768.2 | 4 - 5 | 572 | 157 | ||
GenBank mRNA | AF086631.1 | - | 4 - 5 | 398 | 157 | |
BC065576.1 | - | 4 - 5 | 583 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
